Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat TGN38 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8059
This item is not returnable.
View return policy
Description
TGN38 Polyclonal specifically detects TGN38 in Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
TGN38 | |
Polyclonal | |
Unconjugated | |
P19814 | |
Tgoln2 | |
Human embryonic kidney cell line HEK293-derived recombinant rat TGN38 Leu18-Ser303 Accession # P19814 | |
100 μg | |
Primary | |
Detects rat TGN38 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse TGN38 is observed. | |
Rat | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
TGN46, TGN46TGN38 homolog, TGN48MGC14722, TGN51, TGN5148, 51kD isoforms), Tgoln1, Tgoln2, trans-golgi network protein 2, Trans-Golgi network protein TGN51, Ttgn1 | |
Sheep | |
Affinity Purified | |
RUO | |
10618 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction