Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat TIMP-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF580
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF580 100 μg
AF580SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF580 Supplier R&D Systems Supplier No. AF580
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

TIMP-1 Polyclonal specifically detects TIMP-1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen TIMP-1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P30120
Gene Alias CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Gene Symbols TIMP1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat TIMP-1 Cys24-Ala217 Accession # P30120
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7076
Test Specificity Detects rat TIMP-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) TIMP-1 and recombinant mouse TIMP-1 is observed and less than 1% cross-reactivity with rhTIMP-2, rhTIMP-3, and rhTIMP-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.