Learn More
Rat TNF-alpha Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | TNF-alpha |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Clone | 45418 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P16599 |
| Gene Alias | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Gene Symbols | TNF |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.