Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat TNF-alpha Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF510
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF510 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF510 Supplier R&D Systems Supplier No. BAF510
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

TNF-alpha Polyclonal specifically detects TNF-alpha in Rat samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen TNF-alpha
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P16599
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Goat
Immunogen E. coli-derived recombinant rat TNF-alpha (R&D Systems, Catalog # 510-RT) Leu80-Leu235 Accession # P16599
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects rat TNF-alpha in ELISAs and Western blots. In sandwich immunoassays, less than 2.5% cross-reactivity with rmTNF-a is observed and less than 0.2% cross-reactivity with rhTNF-a and rpTNF-a is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.