Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat TNF-alpha Quantikine ELISA Kit

Catalog No. p-4753971
Click to view available options
Quantity:
2 Plates
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Rat TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat TNF-alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat TNF-alpha.

Specifications

Assay Range 12.50 to 800 pg/mL
Assay Sensitivity 5 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum
Specificity Natural and Recombinant Rat TNF-alpha
Sufficient For 6 x 96-well Plates
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 24835
Quantity 6 Plates
Sample Volume Cell Culture Supernates (17 μL), Serum (25 μL), EDTA Plasma (25 μL), Heparin Plasma (25 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.