Learn More
Description
- This kit recognizes Rat ACP5 in samples.
- No significant cross-reactivity or interference between Rat ACP5 and analogues was observed.
- Assay Type: Sandwich ELISA
- Standard Curve Range: 0.63 - 40 ng/mL
- Sensitivity: 0.38 ng/mL
- Inter-Assay: 4.71%
- Intra-Assay: 4.5%
- Spike Recovery: 84-109%
- Sample Volume: 100 μL
- Components:
- Substrate Reagent
- Reference Standard & Sample Diluent
- Stop Solution
- Reference Standard
- Product Description
- Micro ELISA Plate(Dismountable)
- Plate Sealer
- HRP Conjugate Diluent
- Biotinylated Detection Ab Diluent
- Certificate of Analysis
- Concentrated HRP Conjugate (100×)
- Concentrated Biotinylated Detection Ab (100×)
- Concentrated Wash Buffer (25×)
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Gene Alias | HPAP, TRACP5a, TRACP5b, TRAP, TrATPase |
| Gene ID (Entrez) | 54 |
| Gene Symbol | ACP5 |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
