Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat TSH beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4507
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4507 100 μg
MAB4507SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4507 Supplier R&D Systems Supplier No. MAB4507
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSH beta Monoclonal specifically detects TSH beta in Rat samples. It is validated for Western Blot.

Specifications

Antigen TSH beta
Applications Western Blot
Classification Monoclonal
Clone 442506
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P04652
Gene Alias thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, Thyrotropin beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSHB, TSH-B, TSH-BETA
Gene Symbols TSHB
Host Species Mouse
Immunogen E. coli-derived recombinant rat TSH beta Phe21-Gly138 Accession # P04652
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7252
Test Specificity Detects rat TSH beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 5% cross-reactivity with recombinant human TSH beta .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.