Learn More
Specifications
Specifications
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.09 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat UCP1 in samples. No Significant cross-reactivity or interference between Rat UCP1 and analogues was observed. |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7350 |
| Gene Symbol | UCP1 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
