Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat UNC5H1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1405
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1405 Supplier R&D Systems Supplier No. AF1405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

UNC5H1/Unc5a Polyclonal specifically detects UNC5H1/Unc5a in Rat samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry.

Specifications

Antigen UNC5H1/Unc5a
Applications Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O08721
Gene Alias KIAA1976netrin receptor UNC5A, netrin receptor Unc5h1, Protein unc-5 homolog 1, Protein unc-5 homolog A, unc5 (C.elegans homolog) a, unc-5 homolog 1, unc-5 homolog A (C. elegans), UNC5A, UNC5H1FLJ16449
Gene Symbols UNC5A
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat UNC5H1 Gln26-Asp358 Accession # O08721
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90249
Test Specificity Detects rat UNC5H1 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant rat UNC5H2, recombinant human (rh) UNC5H3 and rhUNC5H4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.