Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat UNC5H1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1405
This item is not returnable.
View return policy
Description
UNC5H1/Unc5a Polyclonal specifically detects UNC5H1/Unc5a in Rat samples. It is validated for Western Blot.Specifications
UNC5H1/Unc5a | |
Polyclonal | |
Biotin | |
O08721 | |
UNC5A | |
Mouse myeloma cell line NS0-derived recombinant rat UNC5H1 Gln26-Asp358 Accession # O08721 | |
50 μg | |
Primary | |
Detects rat UNC5H1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant rat UNC5H2 and recombinant human UNC5H4 is observed. | |
Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
KIAA1976netrin receptor UNC5A, netrin receptor Unc5h1, Protein unc-5 homolog 1, Protein unc-5 homolog A, unc5 (C.elegans homolog) a, unc-5 homolog 1, unc-5 homolog A (C. elegans), UNC5A, UNC5H1FLJ16449 | |
Goat | |
Affinity Purified | |
RUO | |
90249 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction