Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat UNC5H2/UNC5B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1006SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1006SP 25 μg
MAB1006 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1006SP Supplier R&D Systems Supplier No. MAB1006SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

UNC5H2/UNC5B Monoclonal specifically detects UNC5H2/UNC5B in Rat samples. It is validated for Western Blot.

Specifications

Antigen UNC5H2/UNC5B
Applications Western Blot
Classification Monoclonal
Clone 187305
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O08722
Gene Alias p53RDL1, p53-regulated receptor for death and life protein 1, Protein unc-5 homolog 2, Protein unc-5 homolog B, transmembrane receptor Unc5H2, unc5 (C.elegans homolog) b, unc-5 homolog 2, unc-5 homolog B (C. elegans), UNC5B, UNC5H2netrin receptor UNC5B
Gene Symbols UNC5B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373 Accession # O08722
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219699
Test Specificity Detects rat UNC5H2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat UNC5H1 or recombinant human UNC5H4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.