Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat Undercarboxylated Osteocalcin ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Description
No Significant cross-reactivity or interference between Rat ucOC and analogues was observed
Specifications
Specifications
Assay Range | 78.13 - 5000 pg/mL |
Assay Sensitivity | 46.88 pg/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Other Biological Fluids, Plasma, Serum |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 632 |
Host Species | Rat |
Quantity | 1 Kit |
Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Target | Undercarboxylated Osteocalcin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction