Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat Universal Protein Whole Tissue Lysate (Adult Whole Normal)
SDP

Catalog No. NB82089440 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB82089440 2 vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB82089440 Supplier R&D Systems™ Supplier No. NB82089440
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

The Rat Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from rat. It has been validated for the following applications: Western Blot, Immunoprecipitation.

Specifications

Quantity 2 vials
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Universal Protein Lysate: Rat Normal Tissues
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Rat
Host Species Rat
Concentration 5 mg/mL
Product Type Rat Universal Protein Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Product Line Novus
Protein State Native
For Use With (Application) Western Blot
Tissue Universal
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.