Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat VAP-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7329
This item is not returnable.
View return policy
Description
VAP-B Monoclonal specifically detects VAP-B in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VAP-B | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9Z269 | |
VAPB | |
E. coli-derived recombinant rat VAP-B Ala2-Pro132 Accession # Q9Z269 | |
100 μg | |
Primary | |
Detects rat VAP-B in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) VAP-B and approximately 50% cross-reactivity with rhVAP-A is observed. | |
Rat | |
Purified |
Western Blot, Immunohistochemistry | |
699811 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALS8, ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAMP-C, VAPB, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9217 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction