Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat VCAM-1/CD106 ELISA Kit (Colorimetric)
SDP

Catalog No. NB397991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397991 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397991 Supplier Novus Biologicals™ Supplier No. NBP308191
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 12.5 to 800 pg/mL
Assay Sensitivity 7.5 pg/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 7412
Gene Symbol VCAM1
Quantity 1 Kit
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.