Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat/Xenopus Potassium Channel Kv2.2 Antibody, R&D Systems™

Catalog No. PPS015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS015 Supplier R&D Systems Supplier No. PPS015
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Potassium Channel Kv2.2 Polyclonal specifically detects Potassium Channel Kv2.2 in Rat, Xenopus samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Potassium Channel Kv2.2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 dilution
Gene Alias delayed rectifier potassium channel protein, KCNB2, KV2.2, potassium channel Kv2.2, potassium voltage-gated channel subfamily B member 2, potassium voltage-gated channel, Shab-related subfamily, member 2, voltage-gated potassium channel subunit Kv2.2
Gene Symbols KCNB2
Host Species Rabbit
Immunogen Peptide from the Kv2.2 subunit region of Xenopus and rodent, conjugated to keyhole limpet hemocyanin (KLH)
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9312
Test Specificity Rodent and Xenopus ∼125 kDa Kv2.2 subunit of the Voltage Gated Potassium Channel protein.
Target Species Rat, Xenopus species
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.