Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Rattin Antibody, PE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300246PE
Description
Rattin Polyclonal antibody specifically detects Rattin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| Rattin | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| Humanin-like in rat, rattin humanin-like | |
| A synthetic peptide mapping to residues 20-35 [VKRLESPNKTRRPYGA] of the Rattin protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Human, Rat | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            