Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rattin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146257 Supplier Invitrogen™ Supplier No. PA146257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rattin Polyclonal Antibody for Western Blot, IHC (P)

A novel rat gene, Rattin, encodes a peptide homologous to Humanin, a secreted peptide that specifically protects against neuronal cell death induced by beta-amyloid peptide (Ab) or by mutations causing early-onset familial Alzheimer's disease. Rattin is the prototype of a novel class of peptides, phylogenetically related to Humanin, endowed with protective activity not only against Ab but also toward excitotoxic neuronal death. The rat gene, Rattin, encodes a peptide of 38 residues (15 residues longer than Humanin) showing 73% identity in the conserved region to Humanin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rattin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias Humanin homolog
Host Species Rabbit
Immunogen A synthetic peptide mapping to residues 20-35 of the Rattin protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.