Learn More
Restek™ RAVEqc Valve Bracket
SDP
Catalog No. 06611876 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Restek Air Sampling Canisters (1, 3, 6, 15 L)
Catalog No. For Use With (Equipment)
06-611-876 Restek Air Sampling Canisters (1, 3, 6, 15 L)
1 options
Catalog No. 06-611-876 Supplier Restek™ Supplier No. 27375
Required for use with Restek 1, 3, 6, and 15 L Air Sampling Canisters when using the Compression Fitting mounted directly on the 1/4 in. Canister Tube Stub
  • Not for use with Restek™ Miniature Canisters or Entech Canisters
Specifications
Type RAVEqc Valve Bracket
For Use With (Application) Attaching to Restek™ Canister
For Use With (Equipment) Restek Air Sampling Canisters (1, 3, 6, 15 L)