Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAVER1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$501.50
Specifications
Antigen | RAVER1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RAVER1 Polyclonal specifically detects RAVER1 in Human samples. It is validated for Western Blot.Specifications
RAVER1 | |
Polyclonal | |
Rabbit | |
KIAA1978Protein raver-1, ribonucleoprotein PTB-binding 1, ribonucleoprotein, PTB-binding 1 | |
RAVER1 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
125950 | |
Synthetic peptides corresponding to RAVER1(ribonucleoprotein, PTB-binding 1) The peptide sequence was selected from the middle region of RAVER1. Peptide sequence ALLQLALQTQGQKKPGILGDSPLGALQPGAQPANPLLGELPAGGGLPPEL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title