Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAVER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541934 Supplier Invitrogen™ Supplier No. PA541934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VTHRPPLSPK SGAEVEAGDA AERRAPEEEL PPLDPEEIRK RLEHTERQFR Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Rabbit: 86%.

RAVER1 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAVER1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAVER1
Gene Accession No. Q8IY67
Gene Alias 1300006N24Rik; AA050359; Kiaa1978; LOW QUALITY PROTEIN: ribonucleoprotein PTB-binding 1; mKIAA1978; Protein raver-1; PTB-binding 1; RAVER1; Ribonucleoprotein; Ribonucleoprotein PTB-binding 1; ribonucleoprotein, PTB binding 1; ribonucleoprotein, PTB-binding 1
Gene Symbols RAVER1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RAVER1 (aa 7-56).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125950
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.