Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAVER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589015 Supplier Invitrogen™ Supplier No. PA589015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPALSTALLQ LALQTQGQKK PGILGDSPLG ALQPGAQPAN PLLGELPAGG GLPPELPPRR GKPPPLLPSV LGPAGGDREA LGLGPPAAQL TPPPAPVGLR G; Positive Samples: 293T, HeLa; Cellular Location: Cytoplasm, Nucleus.

RAVER1 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAVER1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RAVER1
Gene Accession No. Q8IY67, Q9CW46
Gene Alias 1300006N24Rik; AA050359; Kiaa1978; LOW QUALITY PROTEIN: ribonucleoprotein PTB-binding 1; mKIAA1978; Protein raver-1; PTB-binding 1; RAVER1; Ribonucleoprotein; Ribonucleoprotein PTB-binding 1; ribonucleoprotein, PTB binding 1; ribonucleoprotein, PTB-binding 1
Gene Symbols RAVER1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 390-490 of human RAVER1 (NP_5977092).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125950, 71766
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.