Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAVER2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569101 Supplier Invitrogen™ Supplier No. PA569101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Human: 100%; Mouse: 79%; Rabbit: 92%.

RAVER2 may bind single-stranded nucleic acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAVER2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAVER2
Gene Accession No. Q9HCJ3
Gene Alias A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2
Gene Symbols RAVER2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RAVER2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55225
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.