Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAVER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113389
Description
RAVER2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
RAVER2 may bind single-stranded nucleic acids.
Specifications
| RAVER2 | |
| Polyclonal | |
| Unconjugated | |
| RAVER2 | |
| A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55225 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9HCJ3 | |
| RAVER2 | |
| Recombinant Human Ribonucleoprotein PTB-binding 2 protein (2-691AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction