Learn More
Specifications
Specifications
| Preparation Method | RAW264.7 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard mini-gel blot (1.0 mg/mL). |
| Quantity | 2 Vials |
| Host Species | Mouse |
| Content And Storage | Store at -20°C to -80°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Target | Raw 264.7 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
