Learn More
Eisco™ Raw Chlorite, Metamorphic Mineral Specimen - Approx. 3 Inch
SCI_ED
Catalog No. S02919 Shop All Eisco Products
Change view
Click to view available options
Product Type:
Metamorphic Rock
Catalog No. Product Type
S02919 Metamorphic Rock
1 options
Catalog No. S02919 Supplier Eisco™ Supplier No. ESNG0068HAND
Add to Cart
Add to Cart
Chlorite is a metamorphic mineral that has undergone light alteration. During metasomatism, phyllosilicate minerals may become hydrated and form chlorite.
  • Chlorite sample approximately 3 inch in size
  • Platy metamorphic mineral with a soapy feel
  • Perfect size for use in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements
  • Forms when hot water alters the chemistry of an existing mineral
  • Specimen comes in clear labeled and barcoded bag for ideal storage
Specifications
Product Type Metamorphic Rock