Learn More
Eisco™ Raw Fossiliferous Limestone, Sedimentary Rock Specimen - Approx. 3 Inch
SCI_ED
Catalog No. S02923 Shop All Eisco Products
Change view
Click to view available options
Specimen Size:
3 in.
Catalog No. Specimen Size
S02923 3 in.
1 options
Catalog No. S02923 Supplier Eisco™ Supplier No. ESNG0034HAND
Add to Cart
Add to Cart
Fossiliferous Limestone Specimen for sedimentary rock identification and studying fossils within whole rocks.
  • This rock formed at the bottom of a marine environment through the compaction of deceased organic matter in a clay or silt cement
  • Ideal for identification and classification exercises, for studying fossils within whole rocks and sedimentary rocks
  • Each specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features
  • Our rock and mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Each specimen sample is packaged in a transparent bag, and is labeled and barcoded for easy identification
Specifications
Product Type Sedimentary Rock
Specimen Size 3 in.