Learn More
Eisco™ Raw Quartz, Mineral Specimen - Approx. 3 Inch
SCI_ED
Catalog No. S02925 Shop All Eisco Products
Change view
Click to view available options
Product Type: Mineral
Catalog No.
S02925 Product Type: Mineral
1 options
Catalog No. S02925 Supplier Eisco™ Supplier No. ESNG0010HAND
Only null left
Add to Cart
Add to Cart
Quartz mineral specimen for use in classroom identification labs and exercises.
  • One of the most abundant minerals on Earth. The tightly packed framework of silica tetrahedral that makes up this mineral gives it a high hardness of 7, a lack of cleavage planes, and a distinct euhedral crystal habit
  • A must-have mineral for any specimen collection. Quartz is useful during lessons on mineral hardness, luster, and fracture
  • Each specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features
  • Our rock and mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Each specimen sample is packaged in a transparent bag, and is labeled and barcoded for easy identification
Specifications
Product Type Mineral