Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAX Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB10056467AF350

Catalog No. NB003555


Only null left
Add to Cart

Description

Description

RAX Polyclonal specifically detects RAX in Human, Canine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

RAX
Polyclonal
Western Blot
MCOP3, retina and anterior neural fold homeobox, retinal homeobox protein Rx, RXRetina and anterior neural fold homeobox protein
Rabbit
Affinity Purified
RUO
The amino acid sequence used as immunogen has 100% identity in human, dog, and monkey.
Store at 4°C in the dark.
Western Blot
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
RAX
A portion of amino acids 300-350 of human Rax was used as the immunogen.
0.1 mL
Primary
Human, Canine, Primate
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.