Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAX Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056467APC
Description
RAX Polyclonal antibody specifically detects RAX in Human, Canine, Primate samples. It is validated for Western BlotSpecifications
RAX | |
Polyclonal | |
Western Blot | |
MCOP3, retina and anterior neural fold homeobox, retinal homeobox protein Rx, RXRetina and anterior neural fold homeobox protein | |
A portion of amino acids 300-350 of human Rax was used as the immunogen. | |
0.1 mL | |
Primary | |
Human, Canine, Primate | |
Purified |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
30062 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction