Learn More
RAX Mouse anti-Human, Clone: OTI1F11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810995

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Rax plays a critical role in eye formation by regulating the initial specification of retinal cells and/or their subsequent proliferation. It binds to the photoreceptor conserved element-I (PCE-1/Ret 1) in the photoreceptor cell-specific arrestin promoter.Specifications
RAX | |
Monoclonal | |
Unconjugated | |
RAX | |
MCOP3, RX | |
Mouse | |
Affinity Chromatography | |
RUO | |
30062 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1F11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y2V3 | |
RAX | |
Human recombit protein fragment corresponding to amino acids 1-222 of human RAX produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.