Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540452 Supplier Invitrogen™ Supplier No. PA540452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MHLPGCAPAM ADGSFSLAGH LLRSPGGSTS RLHSIEAILG FTKDDGILGT Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Rax plays a critical role in eye formation by regulating the initial specification of retinal cells and/or their subsequent proliferation. It binds to the photoreceptor conserved element-I (PCE-1/Ret 1) in the photoreceptor cell-specific arrestin promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAX
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAX
Gene Accession No. Q9Y2V3
Gene Alias 7530406A22Rik; E130303K03Rik; ey1; ey-1; eyeless 1; LOW QUALITY PROTEIN: retinal homeobox protein Rx; MCOP3; RAX; retina and anterior neural fold homeobox; retina and anterior neural fold homeobox protein; Retinal homeobox protein Rx; Rx; Rx/rax homeoprotein
Gene Symbols RAX
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RAX (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30062
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.