Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562633 Supplier Invitrogen™ Supplier No. PA562633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERLESGSGAV AAPRLPEAPA LPFARPPAMS LPLEPWLG Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 39%.

This gene encodes a homeodomain-containing protein that plays a role in eye development. Mutation of this gene causes age-related macular degeneration type 6, an eye disorder resulting in accumulations of protein and lipid beneath the retinal pigment epithelium and within the Bruch's membrane. Defects in this gene can also cause cone-rod dystrophy type 11, a disease characterized by the initial degeneration of cone photoreceptor cells and resulting in loss of color vision and visual acuity, followed by the degeneration of rod photoreceptor cells, which progresses to night blindness and the loss of peripheral vision.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAX2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAX2
Gene Accession No. Q96IS3
Gene Alias ARMD6; CORD11; Q50-type retinal homeobox protein; QRX; RAX2; RAXL1; retina and anterior neural fold homeobox 2; retina and anterior neural fold homeobox like 1; retina and anterior neural fold homeobox protein 2; Retina and anterior neural fold homeobox-like protein 1
Gene Symbols RAX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAX2. Recombinant protein control fragment (Product #RP-99912).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.