Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rb (Cleaved DRb-p70) Monoclonal Antibody (172C1094)
GREENER_CHOICE

Catalog No. 01676588
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-588 Supplier Invitrogen™ Supplier No. MA141186
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: camptothecin-treated HL-60 (24 hr).

The retinoblastoma gene product (Rb) is a tumor suppressor protein and acts as cell cycle regulator (1). Recently, Rb has been shown to play a role in apoptosis. During apoptosis, Rb is subjected to cleavage by caspase-3 like protease. Caspase-3 cleaves Rb into 48 and 68kDa fragments (2-4). Cleavage of Rb would make it functionally inactive, causing release of E2F-1. The E2F-1 then may promote apoptosis (5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rb (Cleaved DRb-p70)
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 172C1094
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene RB1
Gene Accession No. P06400, P13405
Gene Alias Cleaved Rb; delta RB-p70; DRb-p70; exon 17 tumor GOS561 substitution mutation causes premature stop; GOS563 exon 17 substitution mutation causes premature stop; OSRC; p105-Rb; pp105; pp110; PPP1R130; pRb; prepro-retinoblastoma-associated protein; protein phosphatase 1, regulatory subunit 130; Rb; RB transcriptional corepressor 1; RB1; Rb-1; Rb-p70; retinoblastoma 1; retinoblastoma suspectibility protein; retinoblastoma-associated protein
Gene Symbols RB1
Host Species Mouse
Immunogen A synthetic peptide corresponding to a region of the caspase-3-induced cleavage site of human Rb.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19645, 5925
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.