Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB1 Antibody (172C1094) - (cleaved), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | cleaved Retinoblastoma 1 |
---|---|
Clone | 172C1094 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Knockdown Validated |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056598T
![]() |
Novus Biologicals
NB10056598SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056598
![]() |
Novus Biologicals
NB10056598 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
RB1 Monoclonal specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.Specifications
cleaved Retinoblastoma 1 | |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors | |
Q03137 | |
5925 | |
A synthetic peptide corresponding to a region of the caspase-3-induced cleavage site of human Rb was used as immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
172C1094 | |
Western Blot 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Knockdown Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
RB1 | |
IgG1 | |
Protein G purified | |
The antibody recognizes cleaved Rb (DRb-p70). The antibody does not recognize full-length Rb. In camptothecin treated HL60, an ∼68 kDa band is observed. DRb-p70 exists in vivo as an N-terminally truncated form of Rb (Liu et al.). DRb-p70 is thought to be the product of alternative translation and is expressed in primary myeloid cells in fetal liver, bone marrow and spleen. DRb-p70 is also specifically expressed in tumor-associated macrophages (TAMs). TAMs are an important targets of cancer therapy. DRb-p70 is a marker of TAMs, and maybe useful for identifying tumors with macrophage infiltration (Liu et al, 2005). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title