Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB1 Antibody (RB1/2313R), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308940UV
Description
RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
RB1 | |
Monoclonal | |
DyLight 350 | |
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
Recombinant human full-length RB1 protein (Uniprot: P06400) | |
100 μg | |
Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors | |
5925 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
RB1/2313R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction