Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Antibody (SPM353), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$235.64 - $529.81

Specifications

Antigen RB1
Clone SPM353
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25339920
SDP
View Documents
Novus Biologicals
NBP25339920UG
20 μg
Each for $235.64
Only null left
Add to Cart
 
NB253399100
SDP
View Documents
Novus Biologicals
NBP253399100UG
100 μg
Each for $529.81
Only null left
Add to Cart
 
NB253399200
SDP
View Documents
Novus Biologicals
NBP253399200UG
200 μg N/A N/A N/A
Description

Description

RB1 Monoclonal specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

RB1
0.2 mg/mL
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
5925
Recombinant human Rb1 protein fragment of 283 amino acid residues (exact sequence is proprietary)
Primary
Store at 4C.
SPM353
Western Blot 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
RB1
IgG1 κ
Protein A or G purified
105 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.