Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Mouse anti-Human, Clone: SPM353, Abnova™

Catalog No. 89034757 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-757 Supplier Abnova Corporation Supplier No. MAB14674
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human RB1.

  • High specificity
  • High reactivity

Specifications

Antigen RB1
Applications Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SPM353
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human RB1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 - 2 ug/mL) Immunoprecipitation (0.5 - 1 ug/500 ug protein lysate) Western Blot (0.5 - 1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene Accession No. P06400
Gene Alias OSRC/RB/p105-Rb/pRb/pp110
Gene Symbols RB1
Host Species Mouse
Immunogen Recombinant protein corresponding to a 283 amino acid fragment of human RB1.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5925
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.