Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB1 Mouse anti-Human, Mouse, Clone-SPM353, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254477200UG
Description
IHC-P:Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10mM Tris with 1mM EDTA Buffer pH 9.0 for 10-20 min followed by cooling at RT for 20 minutesSpecifications
RB1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
Mouse | |
105 kDa | |
200 μg | |
Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors | |
5925 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
SPM353 | |
Unconjugated | |
PBS with No Preservative | |
RB1 | |
Recombinant human Rb1 protein fragment of 283 amino acid residues (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction