Learn More
RB1 (phospho S780) Rabbit anti-Human, Mouse, Rabbit, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human RB1.
Supplier: Abnova Corporation PAB29613
Description
The protein encoded by this gene is a negative regulator of the cell cycle and was the first tumor suppressor gene found. The encoded protein also stabilizes constitutive heterochromatin to maintain the overall chromatin structure. The active, hypophosphorylated form of the protein binds transcription factor E2F1. Defects in this gene are a cause of childhood cancer retinoblastoma (RB), bladder cancer, and osteogenic sarcoma. [provided by RefSeq]
Specifications
RB1 (phospho S780) | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human RB1. | |
In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
RB1 | |
A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S780 of human RB1. | |
100 μL | |
Primary | |
RB1 (phospho S780) polyclonal antibody detects endogenous level of RB1 only when phosphorylated at serine 780. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1:100-1:200) Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
OSRC/RB/p105-Rb/pRb/pp110 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5925 | |
Human, Mouse, Rabbit | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.