Learn More
Rb1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302432AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.Specifications
Rb1 | |
Polyclonal | |
Unconjugated | |
RB1 | |
Phospho-Retinoblastoma, OSRC, P105RB, PP105, PP110, pRb, Retinoblastoma, Retinoblastoma associated protein, Retinoblastoma related osteosarcoma, Retin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5925 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P06400 | |
RB1 | |
Between 550 and 600 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.