Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM15B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557749 Supplier Invitrogen™ Supplier No. PA557749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKKARDSERN HRTTEAEPKP LEEPKHETKK LKNLSEYAQT LQLGWNGLLV LKNSCFPTSM HILEGDQGVI SSLLKDHTSG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

May function in the regulation of alternative or illicit splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM15B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBM15B
Gene Accession No. Q8NDT2
Gene Alias 1810017N16Rik; chromosome 3p21.1 gene sequence; HsOTT3; HUMAGCGB; huOTT3; one twenty two protein 3; one-twenty two protein 3; OTT3; putative RNA-binding protein 15B; Rbm15b; RGD1305937; RNA binding motif protein 15 pseudogene; RNA binding motif protein 15B; rna-binding; RNA-binding motif protein 15B
Gene Symbols RBM15B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RB15B. Recombinant protein control fragment (Product #RP-96501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.