Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM15B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566136
Description
Immunogen sequence: LHGGYQYKQRS LSPVAAPPLR EPRARHAAAA FALDAAAAAA VGLSRERALD YYGLYDDRGR PYGYPAVCEE DLMPEDDQRA T Highest antigen sequence identity to the following orthologs - mouse 99%, rat 98%.
May function in the regulation of alternative or illicit splicing.
Specifications
RBM15B | |
Polyclonal | |
Unconjugated | |
RBM15B | |
1810017N16Rik; chromosome 3p21.1 gene sequence; HsOTT3; HUMAGCGB; huOTT3; one twenty two protein 3; one-twenty two protein 3; OTT3; putative RNA-binding protein 15B; Rbm15b; RGD1305937; RNA binding motif protein 15 pseudogene; RNA binding motif protein 15B; rna-binding; RNA-binding motif protein 15B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29890 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDT2 | |
RBM15B | |
Recombinant Human RB15B. Recombinant protein control fragment (Product #RP-105092). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction