Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM15B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569092
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%.
May function in the regulation of alternative or illicit splicing.
Specifications
RBM15B | |
Polyclonal | |
Unconjugated | |
RBM15B | |
1810017N16Rik; chromosome 3p21.1 gene sequence; HsOTT3; HUMAGCGB; huOTT3; one twenty two protein 3; one-twenty two protein 3; OTT3; putative RNA-binding protein 15B; Rbm15b; RGD1305937; RNA binding motif protein 15 pseudogene; RNA binding motif protein 15B; rna-binding; RNA-binding motif protein 15B | |
Rabbit | |
Affinity chromatography | |
RUO | |
109095 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6PHZ5 | |
RBM15B | |
synthetic peptide directed towards the middle region of human RB15B (aa 606-655). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction