Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM15B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110279
Description
Immunogen sequence: DRDRTFLEGD WTSPSKSSDR RNSLEGYSRS VRSRSGERWG ADGDRGLPKP WEERRKRRSL SSDRGRTTHS PYEERSRTKG SGQQSERGSD RTPERSRKEN HSSEGTKESS SNSLSNSRHG AEERGHHHHH HEAADSSHGK KARDSERNHR TTEAEPKPLE EPKHETKKLK NLSEYAQTLQ L.
May function in the regulation of alternative or illicit splicing.
Specifications
RBM15B | |
Polyclonal | |
Unconjugated | |
RBM15B | |
1810017N16Rik; chromosome 3p21.1 gene sequence; HsOTT3; HUMAGCGB; huOTT3; one twenty two protein 3; one-twenty two protein 3; OTT3; putative RNA-binding protein 15B; Rbm15b; RGD1305937; RNA binding motif protein 15 pseudogene; RNA binding motif protein 15B; rna-binding; RNA-binding motif protein 15B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
109095, 29890 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.91 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6PHZ5, Q8NDT2 | |
RBM15B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 540-720 of human RBM15B (NP_037418.3). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction