Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB1CC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077317
Description
RB1CC1 Polyclonal specifically detects RB1CC1 in Human samples. It is validated for Immunoprecipitation.Specifications
RB1CC1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
CC1, DRAGOU14, FIP200, KIAA0203200 kDa FAK family kinase-interacting protein, RB1-inducible coiled-coil 1, RB1-inducible coiled-coil protein 1, RBICC | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 1:10-1:500 | |
NP_055596.2 | |
RB1CC1 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 human 200 kDa FAK family kinase-interacting protein (RB1-inducible coiled-coil 1) using the numbering given in entry NP_055596.2 (GeneID 9821). | |
100 μL | |
Tumor Suppressors | |
9821 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction