Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB545 Hamster Anti-Mouse CD11c (Integrin αX), BD Horizon™

Catalog No. BDB569775 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569775 25 μg
BDB569747 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569775 Supplier BD Biosciences Supplier No. 569775
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

CD11c (Integrin αX) Monoclonal antibody specifically detects CD11c (Integrin αX) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c (Integrin αX)
Applications Flow Cytometry
Classification Monoclonal
Clone N418
Concentration 0.2 mg/mL
Conjugate RB545
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4
Host Species Hamster
Immunogen Mouse Dendritic Cells
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.