Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB545 Hamster Anti-Mouse CD3e, BD Horizon™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 569753
Description
CD3e Monoclonal antibody specifically detects CD3e in Mouse samples. It is validated for Flow cytometry
Specifications
CD3e | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
145-2C11 | |
RB545 | |
CD3; CD3 epsilon; Cd3e; CD3ε; T3e | |
H-2Kb specific cytotoxic T lymphocyte clone BM10-37 | |
25 μg | |
12501 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction