Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB545 Mouse Anti-Human CD27, BD Horizon™
Mouse Monoclonal Antibody
$162.00 - $396.00
Specifications
Antigen | CD27 |
---|---|
Clone | O323 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB545 |
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow cytometrySpecifications
CD27 | |
Flow Cytometry | |
RB545 | |
Mouse | |
Human | |
CD27 antigen; TNFRSF7; S152; T14; Tp55 | |
Human Thymocytes | |
Affinity Chromatography |
O323 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
939 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title