Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB545 Mouse Anti-Human CD86 (B7-2), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569762
Description
CD86 (B7-2) Monoclonal antibody specifically detects CD86 (B7-2) in Human samples. It is validated for Flow cytometry
Specifications
CD86 (B7-2) | |
Monoclonal | |
RB545 | |
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; CD28LG2; LAB72 | |
Human ARH 77 Cell Line | |
25 Tests | |
942 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
BU63 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction